Yttrium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11185-06
11185-14
11185-22
11185-30
CAAA11185-06EA
53.25
CAD
CAAA11185-06
CAAA11185-14
CAAA11185-22
CAAA11185-30
Yttrium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium (III) chloride hydrate
MDL: MFCD00149941
Purity: 99.99%
CAS No.: 12741-05-8
Molecular Formula: Cl3Y.xH2O
Formula Weight: 195.26(anhy)
Physical Form: Crystalline Aggregates
Density: 2.18
Appearance: White to pink
Odor: Odorless
Merck Reference: 14,10107
MDL No.: MFCD00149941
Formula:
Cl₃Y·H₂O MW: 213.28 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149941 CAS Number: 12741-05-8 EINECS: 233-801-0 Merck Index: 14,10107 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...