Diyttrium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Yttrium oxide, Yttrium (III) oxide

11181-14 11181-22 11181-36 11181-A1
CAAA11181-14EA 83.11 CAD
CAAA11181-14 CAAA11181-22 CAAA11181-36 CAAA11181-A1
Diyttrium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Diyttrium trioxide

MDL: MFCD00011473


Purity: 99.99% CAS No.: 1314-36-9 Molecular Formula: O3Y2 Formula Weight: 225.81 Physical Form: 10 Micron Powder Density: 5.01 Appearance: White Odor: Odorless Melting Point: 2410° Merck Reference: 14,10107 MDL No.: MFCD00011473

Formula: Y₂O₃
MW: 225.81 g/mol
Boiling Pt: 4300
Melting Pt: 2410 °C
Density: 5.01
Storage Temperature: Ambient
MDL Number: MFCD00011473
CAS Number: 1314-36-9
EINECS: 215-233-5
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR