Yttrium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger

11178-14 11178-22 11178-36
CAAA11178-14EA 195.92 CAD
CAAA11178-14 CAAA11178-22 CAAA11178-36
Yttrium(III) fluoride, anhydrous ≥99.99% (REO, rare earth oxide basis), REacton®
Yttrium (III) fluoride

Anhydrous


Purity: 99.99% CAS No.: 13709-49-4 Molecular Formula: YF3 Formula Weight: 145.90 Physical Form: Lump/powder Sensitivity: Hygroscopic UN No.: 3288 MDL No.: MFCD00049714

Formula: F₃Y
Boiling Pt: 2227 °C (1013 hPa)
Melting Pt: 1387 °C
Density: 4.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00049714
CAS Number: 13709-49-4
EINECS: 237-257-5
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Form Lump/powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR