Germanium(IV) oxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Germanium dioxide
11155-06
11155-14
11155-22
CAAA11155-06EA
109.81
CAD
CAAA11155-06
CAAA11155-14
CAAA11155-22
Germanium(IV) oxide ≥99.999% (metals basis), Puratronic®
Germanium (IV) oxide
MDL: MFCD00011030
Slightly soluble in sodium hydroxide. Insoluble in water and hydrochloric acid
Purity: 99.999%
CAS No.: 1310-53-8
Molecular Formula: GeO2
Formula Weight: 104.59
Physical Form: Powder
Density: 6.239
Appearance: White
Odor: Odorless
Melting Point: 1086°
Solubility: Slightly soluble in sodium hydroxide. Insoluble in water and hydrochloric acid
Merck Reference: 14,4409
MDL No.: MFCD00011030
Formula:
GeO₂ MW: 104.61 g/mol Melting Pt: 1110 Density: 6.239 Storage Temperature: Ambient |
MDL Number:
MFCD00011030 CAS Number: 1310-53-8 EINECS: 215-180-8 Merck Index: 14,4409 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...