Digallium trioxide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
11151-06
11151-14
11151-22
11151-36
CAAA11151-06EA
116.92
CAD
CAAA11151-06
CAAA11151-14
CAAA11151-22
CAAA11151-36
Digallium trioxide ≥99.999% (metals basis), Puratronic®
Digallium trioxide
MDL: MFCD00011020
Purity: 99.999%
CAS No.: 12024-21-4
Molecular Formula: Ga2O3
Formula Weight: 187.44
Physical Form: -50 Mesh Powder
Density: 6.44
Refractive Index: 1.92
Odor: Odorless
Melting Point: 1900°
Merck Reference: 14,4346
MDL No.: MFCD00011020
Formula:
Ga₂O₃ Melting Pt: 1900 °C Density: 6.44 Storage Temperature: Ambient |
MDL Number:
MFCD00011020 CAS Number: 12024-21-4 EINECS: 234-691-7 Merck Index: 14,4346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...