Gallium trinitrate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

11150-14 11150-22 11150-36
CAAA11150-14EA 488.22 CAD
CAAA11150-14 CAAA11150-22 CAAA11150-36
Gallium trinitrate hydrate ≥99.999% (metals basis), Puratronic®
Gallium trinitrate hydrate

MDL: MFCD00149723


Purity: 99.999% CAS No.: 69365-72-6 Molecular Formula: Ga(NO3)3.xH2O Formula Weight: 255.73(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White Melting Point: 110° dec. Merck Reference: 14,4346 UN No.: 1477 MDL No.: MFCD00149723

Formula: GaN₃O₉·H₂O
Melting Pt: 110 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149723
CAS Number: 69365-72-6
EINECS: 236-815-5
UN: 1477
ADR: 5.1,III
Merck Index: 14,4346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR