Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)

Supplier: Thermo Scientific Chemicals

11144-06 11144-14 11144-22
CAAA11144-06EA 192.76 CAD
CAAA11144-06 CAAA11144-14 CAAA11144-22
Germanium ≥99.9999% (metals basis), pieces, Puratronic® ≤ 2 cm (0.8 in)
Germanium

MDL: MFCD00085310


Purity: 99.9999+% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odor: Odorless MDL No.: MFCD00085310

Formula: Ge
Boiling Pt: 2830 °C (1013 hPa)
Melting Pt: 937.2 °C
Density: 5.323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR