Zinc(II) nitrate hexahydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Zinc nitrate hexahydrate
11136-18
11136-30
CAAA11136-18EA
448.72
CAD
CAAA11136-18
CAAA11136-30
Zinc(II) nitrate hexahydrate ≥99.998% (metals basis), Puratronic®
Zinc (II) nitrate hexahydrate
MDL: MFCD00149889
Purity: 99.998%
CAS No.: 10196-18-6
Molecular Formula: N2O6Zn.6H2O
Formula Weight: 297.47 (189.38anhy)
Physical Form: Crystalline Aggregates
Density: 2.07
Appearance: White
Odor: Odorless
Melting Point: ca 36°
Merck Reference: 14,10144
UN No.: 1514
MDL No.: MFCD00149889
Formula:
ZnN₂O₆·6H₂O Boiling Pt: ∼105 °C (1013 hPa) Melting Pt: ∼36 °C Density: 2.07 Storage Temperature: Ambient |
MDL Number:
MFCD00149889 CAS Number: 10196-18-6 EINECS: 231-943-8 UN: 1514 ADR: 5.1,II Merck Index: 14,10144 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...