Barium acetate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Barium di(acetate), Acetic acid barium salt
11126-14
11126-22
CAAA11126-14EA
431.34
CAD
CAAA11126-14
CAAA11126-22
Barium acetate ≥99.999% (metals basis), Puratronic®
Barium acetate
MDL: MFCD00012447
Purity: 99.999%
CAS No.: 543-80-6
Molecular Formula: C4H6BaO4
Formula Weight: 255.43
Physical Form: Crystalline
Density: 2.47
Appearance: White
Odor: Odorless
Beilstein Reference: 3693411
Merck Reference: 14,965
UN No.: 1564
MDL No.: MFCD00012447
Formula:
(H₃CCOO)₂Ba Melting Pt: ∼450 °C Density: 2.47 Storage Temperature: Ambient |
MDL Number:
MFCD00012447 CAS Number: 543-80-6 EINECS: 208-849-0 UN: 1564 ADR: 6.1,III Merck Index: 14,965 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...