Barium chromate ≥99.995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

11124-06 11124-14
CAAA11124-06EA 93.06 CAD
CAAA11124-06 CAAA11124-14
Barium chromate ≥99.995% (metals basis), Puratronic®
Barium chromate

MDL: MFCD00014180


Purity: 99.998% CAS No.: 10294-40-3 Molecular Formula: BaCrO4 Formula Weight: 253.33 Physical Form: Powder Density: 4.50 Appearance: Yellow Odor: Odorless Merck Reference: 14,972 UN No.: 1479 MDL No.: MFCD00014180

Formula: BaCrO₄
Melting Pt: 1400 °C
Density: 4.50
Storage Temperature: Ambient
MDL Number: MFCD00014180
CAS Number: 10294-40-3
EINECS: 233-660-5
UN: 1479
ADR: 5.1,III
Merck Index: 14,972

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR