Bismuth(III) nitrate oxide ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Bismuth (III) oxide nitrate, (Nitrooxy)oxobismuthine, Bismuth (III) oxynitrate

11122-09 11122-18
CAAA11122-09EA 161.16 CAD
CAAA11122-09 CAAA11122-18
Bismuth(III) nitrate oxide ≥99.998% (metals basis), Puratronic®
Bismuth (III) nitrate oxide

MDL: MFCD00064844


Purity: 99.998% CAS No.: 10361-46-3 Molecular Formula: BiONO3 Formula Weight: 286.99 Physical Form: Powder UN No.: 1477 MDL No.: MFCD00064844

Formula: BiNO₄
MW: 286.98
Density: 4.93 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064844
CAS Number: 10361-46-3
EINECS: 233-792-3
UN: 1477
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR