Calcium iodide hydrate ≥99.9965% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11117-06
11117-14
CAAA11117-06EA
102.38
CAD
CAAA11117-06
CAAA11117-14
Calcium iodide hydrate ≥99.9965% (metals basis), Puratronic®
Calcium iodide hydrate
MDL: MFCD00149622
Freely soluble in water and alcohol
Purity: 99.9965%
CAS No.: 71626-98-7
Molecular Formula: CaI2.xH2O (x≈6)
Formula Weight: 293.89(anhy)
Physical Form: Crystalline
Density: 2.55
Appearance: Yellow
Melting Point: 42° dec.
Solubility: Freely soluble in water and alcohol
Merck Reference: 14,1677
MDL No.: MFCD00149622
Formula:
CaI₂·H₂O Melting Pt: 42 °C (dec.) Density: 2.55 Storage Temperature: Ambient |
MDL Number:
MFCD00149622 CAS Number: 71626-98-7 EINECS: 233-276-8 Merck Index: 14,1677 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...