Calcium sulfate, anhydrous ≥99.993% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11116-04
11116-09
11116-18
CAAA11116-04EA
73.15
CAD
CAAA11116-04
CAAA11116-09
CAAA11116-18
Calcium sulfate, anhydrous ≥99.993% (metals basis), Puratronic®
Calcium sulphate
MDL: MFCD00010912
Purity: 99.993%
CAS No.: 7778-18-9
Molecular Formula: CaO4S
Formula Weight: 136.14
Physical Form: Powder
Density: 2.960
Appearance: White
Odor: Odorless
Melting Point: 1450°
Merck Reference: 14,1706
MDL No.: MFCD00010912
Formula:
CaO₄S Melting Pt: 1450 °C Density: 2.960 Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 EINECS: 231-900-3 Merck Index: 14,1706 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...