Aluminium bromide ≥98%

Supplier: Thermo Scientific Chemicals
Danger

11113-09 11113-22 11113-59
CAAA11113-09EA 42.82 CAD
CAAA11113-09 CAAA11113-22 CAAA11113-59
Aluminium bromide ≥98%
Aluminium bromide

Alkylation, bromination and isomerization catalyst in organic synthesis


Purity: 98% CAS No.: 7727-15-3 Molecular Formula: AlBr3 Formula Weight: 266.71 Physical Form: Crystalline Density: 3.21 Appearance: White to orange Odor: Pungent Melting Point: 97.5° Boiling Point: 265° Solubility: Reacts with water violently, soluble in alcohol, carbon disulfide, ether Merck Reference: 14,332 UN No.: 1725 MDL No.: MFCD00003421


Alkylation, bromination and isomerization catalyst in organic synthesis

Formula: AlBr₃
Boiling Pt: 265 °C
Melting Pt: 97.5 °C
Density: 3.21
Storage Temperature: Ambient
MDL Number: MFCD00003421
CAS Number: 7727-15-3
EINECS: 231-779-7
UN: 1725
ADR: 8,II
Merck Index: 14,332

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR