Cesium sulfate 99,997 Metals basis, Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

11112-09 11112-18
CAAA11112-09EA 135.88 CAD
CAAA11112-09 CAAA11112-18
Cesium sulfate 99,997 Metals basis, Puratronic®
Cesium sulphate

MDL: MFCD00010959


Purity: 99.997% CAS No.: 10294-54-9 Molecular Formula: Cs2O4S Formula Weight: 361.87 Physical Form: Powder Density: 4.243 Appearance: White Odor: Odorless Melting Point: 1010° Merck Reference: 14,2016 MDL No.: MFCD00010959

Formula: Cs₂O₄S
MW: 361.87 g/mol
Boiling Pt: 1627
Melting Pt: 1010
Density: 4.243
Storage Temperature: Ambient
MDL Number: MFCD00010959
CAS Number: 10294-54-9
EINECS: 233-662-6
Merck Index: 14,2016

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR