Rubidium carbonate (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
11105-03
11105-06
11105-14
CAAA11105-03EA
72.84
CAD
CAAA11105-03
CAAA11105-06
CAAA11105-14
Rubidium carbonate (metals basis), Puratronic®
Rubidium carbonate
MDL: MFCD00011188
Purity: 99.975%
CAS No.: 584-09-8
Molecular Formula: CO3Rb2
Formula Weight: 230.95
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 837°
MDL No.: MFCD00011188
Formula:
CO₃Rb₂ Melting Pt: 837 °C Density: 2,43 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011188 CAS Number: 584-09-8 EINECS: 209-530-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...