Sodium metavanadate (≥38% V) ∼96% V ≥ 38%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11092-14
11092-30
11092-A1
CAAA11092-14EA
85.64
CAD
CAAA11092-14
CAAA11092-30
CAAA11092-A1
Sodium metavanadate (≥38% V) ∼96% V ≥ 38%
Sodium metavanadate
MDL: MFCD00011125
Purity: 96%
CAS No.: 13718-26-8
Molecular Formula: NaO3V
Formula Weight: 121.93
Physical Form: Chunks/Powder
Appearance: White to cream
Odor: Odorless
Melting Point: 630°
Merck Reference: 14,8700
UN No.: 3285
MDL No.: MFCD00011125
Formula:
NaO₃V Melting Pt: 630 °C Density: 5.15 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011125 CAS Number: 13718-26-8 EINECS: 237-272-7 UN: 3285 ADR: 6.1,III Merck Index: 14,8700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...