Ammonium metavanadate ≥99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11091-30
11091-A1
CAAA11091-30EA
167.48
CAD
CAAA11091-30
CAAA11091-A1
Ammonium metavanadate ≥99% (metals basis)
Ammonium metavanadate
MDL: MFCD00011430
Purity: 99%
CAS No.: 7803-55-6
Molecular Formula: NH4VO3
Formula Weight: 116.98
Physical Form: Powder
Density: 2.33
Melting Point: 200° dec.
Merck Reference: 14,568
UN No.: 2859
MDL No.: MFCD00011430
Formula:
NH₄VO₃ Melting Pt: 200 °C (dec.) Density: 2.33 Storage Temperature: Ambient |
MDL Number:
MFCD00011430 CAS Number: 7803-55-6 EINECS: 232-261-3 UN: 2859 ADR: 6.1,II Merck Index: 14,568 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...