Vanadyl sulfate hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Vanadium (IV) sulphate oxide hydrate, Vanadium(IV) oxide sulphate hydrate, Vanadium oxide sulphate hydrate, Vanadium oxide sulfate hydrate, Basic vanadium (IV) sulfate hydrate, Basic vanadium (IV) sulphate hydrate, Vanadium (IV) sulfate oxide hydrate, Vanadium(IV) oxide sulfate hydrate

11090-09 11090-18 11090-30
CAAA11090-09EA 69.05 CAD
CAAA11090-09 CAAA11090-18 CAAA11090-30
Vanadyl sulfate hydrate ≥99.9% (metals basis)
Vanadyl sulphate hydrate

MDL: MFCD00149933 Soluble in water


Purity: 99.9% CAS No.: 123334-20-3 Molecular Formula: VOSO4.xH2O Formula Weight: 163.00(anhy) Physical Form: Lump Density: 2.5 Appearance: Blue Odor: Odorless Solubility: Soluble in water Merck Reference: 14,9926 UN No.: 2931 MDL No.: MFCD00149933

Formula: O₅SV·H₂O
MW: 163.00 g/mol (anhy)
Density: 2.5
Storage Temperature: Ambient
MDL Number: MFCD00149933
CAS Number: 123334-20-3
EINECS: 248-652-7
UN: 2931
ADR: 6.1 II
Merck Index: 14,9926

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR