Boron nitride ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

11078-18 11078-30
CAAA11078-18EA 169.06 CAD
CAAA11078-18 CAAA11078-30
Boron nitride ≥99.5% (metals basis)
Boron nitride

MDL: MFCD00011317


Purity: 99.5% CAS No.: 10043-11-5 Molecular Formula: BN (hexagonal) Formula Weight: 24.82 Physical Form: -325 Mesh Powder Appearance: White Odor: Odorless Merck Reference: 14,1346 MDL No.: MFCD00011317

Formula: BN
Melting Pt: 2527 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011317
CAS Number: 10043-11-5
EINECS: 233-136-6
Merck Index: 14,1346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR