Tellurium ≥99.999% (metals basis), powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
11075-09
11075-18
11075-30
CAAA11075-09EA
70.94
CAD
CAAA11075-09
CAAA11075-18
CAAA11075-30
Tellurium ≥99.999% (metals basis), powder -200 mesh
Tellurium
MDL: MFCD00134062
Purity: 99.999%
CAS No.: 13494-80-9
Physical Form: Powder
Appearance: Grey
Odor: Odorless
UN No.: 3288
MDL No.: MFCD00134062
Formula:
Te Boiling Pt: 990 °C (1013 hPa) Melting Pt: 450 °C Density: 8.83 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134062 CAS Number: 13494-80-9 EINECS: 236-813-4 UN: 3288 ADR: 6.1,III Merck Index: 12,09273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...