Tellurium ≥99.99% (metals basis), broken ingot

Supplier: Thermo Scientific Chemicals
Danger

11073-18 11073-30
CAAA11073-18EA 175.38 CAD
CAAA11073-18 CAAA11073-30
Tellurium ≥99.99% (metals basis), broken ingot
Tellurium

MDL: MFCD00134062


Purity: 99.99% CAS No.: 13494-80-9 Odor: Odorless MDL No.: MFCD00134062

Formula: Te
Boiling Pt: 990 °C (1013 hPa)
Melting Pt: 450 °C
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1,III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR