Copper ≥99.5% (metals basis), spherical powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

11070-30 11070-A1 11070-D9
CAAA11070-30EA 79 CAD
CAAA11070-30 CAAA11070-A1 CAAA11070-D9
Copper ≥99.5% (metals basis), spherical powder -100 mesh
Copper

MDL: MFCD00010965


Purity: 99.5% CAS No.: 7440-50-8 Appearance: Copper Odor: Odorless UN No.: 3077 MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III
Merck Index: 13,02545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR