Platinium(IV) oxide hydrate ≥99.9% (metals basis) Pt 71-75%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11049-03
11049-06
CAAA11049-03EA
361.82
CAD
CAAA11049-03
CAAA11049-06
Platinium(IV) oxide hydrate ≥99.9% (metals basis) Pt 71-75%
Platinium (IV) oxide hydrate
MDL: MFCD00066964
Fieser: 1,890 2,332 15,268 21,353
Purity: 99.9%
CAS No.: 52785-06-5
Molecular Formula: O2Pt.xH2O
Formula Weight: 227.09(anhy)
Physical Form: Powder
Appearance: Dark brown
Merck Reference: 14,7527
UN No.: 1479
MDL No.: MFCD00066964
Formula:
O₂Pt·H₂O Melting Pt: 450 °C Density: 10.2 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00066964 CAS Number: 52785-06-5 EINECS: 215-223-0 UN: 1479 ADR: 5.1,II Merck Index: 14,7527 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...