Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis) Pt ≥ 51%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium tetrachloroplatinate(II), Ammonium tetrachloroplatinate

11046-03 11046-06
CAAA11046-03EA 260.7 CAD
CAAA11046-03 CAAA11046-06
Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis) Pt ≥ 51%
Diammonium tetrachloroplatinate

MDL: MFCD00010885


Purity: 99.9% CAS No.: 13820-41-2 Molecular Formula: H8Cl4N2Pt Formula Weight: 372.98 Physical Form: Crystalline Density: 2.936 Appearance: Dark red Odor: Odorless Melting Point: 145° dec. UN No.: 3288 MDL No.: MFCD00010885

Formula: H₈Cl₄N₂Pt
MW: 372.97 g/mol
Melting Pt: 145 °C (dec.)
Density: 2.936
Storage Temperature: Ambient
MDL Number: MFCD00010885
CAS Number: 13820-41-2
EINECS: 237-499-1
UN: 3288
ADR: 6.1,III
Merck Index: 12,00583

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR