Platinum(IV) chloride (≥57% Pt) ≥99.9% (metals basis) Pt ≥ 57%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Platinum tetrachloride

11045-03 11045-06
CAAA11045-03EA 319.16 CAD
CAAA11045-03 CAAA11045-06
Platinum(IV) chloride (≥57% Pt) ≥99.9% (metals basis) Pt ≥ 57%
Platinum (IV) chloride

MDL: MFCD00011182


Purity: 99.9% CAS No.: 13454-96-1 Molecular Formula: PtCl4 Formula Weight: 336.90 Physical Form: Crystalline Density: 4.303 Appearance: Brown Odor: Odorless Melting Point: 370° dec. UN No.: 2923 MDL No.: MFCD00011182

Formula: Cl₄Pt
MW: 336.89 g/mol
Melting Pt: 370 °C (dec.)
Density: 4.303
Storage Temperature: Ambient
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR