Platinum(II) chloride ≥99.9% (metals basis) Pt ≥ 73%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11044-03
11044-06
CAAA11044-03EA
447.14
CAD
CAAA11044-03
CAAA11044-06
Platinum(II) chloride ≥99.9% (metals basis) Pt ≥ 73%
Platinum (II) chloride
MDL: MFCD00011181
Insoluble in water, alcohol, ether. Soluble in HCl2 NH4OH
Purity: 99.9%
CAS No.: 10025-65-7
Molecular Formula: Cl2Pt
Formula Weight: 266.00
Physical Form: Crystalline
Density: 5.87
Appearance: Green-brown
Melting Point: 581° dec.
Solubility: Insoluble in water, alcohol, ether. Soluble in HCl, NH4OH
Merck Reference: 14,7528
UN No.: 3261
MDL No.: MFCD00011181
Formula:
Cl₂Pt Melting Pt: 581 °C (dec.) Density: 5.87 Storage Temperature: Ambient |
MDL Number:
MFCD00011181 CAS Number: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8,III Merck Index: 14,7528 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...