Ruthenium(III) chloride hydrate (≥38% Ru) ≥99.9% (PGM basis) Ru ≥ 38%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11043-04
11043-09
11043-18
CAAA11043-04EA
180.12
CAD
CAAA11043-04
CAAA11043-09
CAAA11043-18
Ruthenium(III) chloride hydrate (≥38% Ru) ≥99.9% (PGM basis) Ru ≥ 38%
Ruthenium (III) chloride hydrate
Oxidation
Purity: 99.9%
CAS No.: 14898-67-0
Molecular Formula: Cl3Ru.xH2O
Formula Weight: 207.43(anhy)
Physical Form: Crystalline Soluble
Sensitivity: Hygroscopic
Appearance: Black
Melting Point: 100°dec
Solubility: Very soluble in water. Soluble in alcohol, acetone
Merck Reference: 14,8302
UN No.: 3260
MDL No.: MFCD00149844
Oxidation
Formula:
Cl₃Ru·xH₂O Melting Pt: 100 °C (dec.) Density: 3.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149844 CAS Number: 14898-67-0 EINECS: 233-167-5 UN: 3260 ADR: 8,III Merck Index: 14,8302 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...