Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis) Pd 29%
Supplier: Thermo Scientific Chemicals
|
Danger
|
11042-04
11042-09
CAAA11042-04EA
301.78
CAD
CAAA11042-04
CAAA11042-09
Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis) Pd 29%
Ammonium hexachloropalladate(IV)
MDL: MFCD00015959
Purity: 99.9%
CAS No.: 19168-23-1
Molecular Formula: (NH4)2PdCl6
Formula Weight: 355.20
Physical Form: Crystalline
Density: 2.42
Appearance: Red-orange
Odor: Odorless
UN No.: 3077
MDL No.: MFCD00015959
Formula:
H₈Cl₆N₂Pd Density: 2,42 Storage Temperature: Ambient |
MDL Number:
MFCD00015959 CAS Number: 19168-23-1 EINECS: 242-854-9 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...