Palladium(II) oxide monohydrate ≥99.9% (metals basis) Pd ≥ 73%
Supplier: Thermo Scientific Chemicals
11041-04
11041-09
CAAA11041-04EA
908.5
CAD
CAAA11041-04
CAAA11041-09
Palladium(II) oxide monohydrate ≥99.9% (metals basis) Pd ≥ 73%
Palladium (II) oxide monohydrate
MDL: MFCD00011172
Purity: 99.9%
CAS No.: 64109-12-2
Molecular Formula: PdO.H2O
Formula Weight: 140.41 (122.40anhy)
Physical Form: Powder
Melting Point: dec.
MDL No.: MFCD00011172
Formula:
OPd·1H₂O Melting Pt: Dec. Storage Temperature: Ambient |
MDL Number:
MFCD00011172 CAS Number: 64109-12-2 EINECS: 215-218-3 Merck Index: 14,6993 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...