Palladium(II) oxide, anhydrous ≥99.9% (metals basis) Pd ≥ 85%

Supplier: Thermo Scientific Chemicals
Warning

11040-03 11040-06
CAAA11040-03EA 455.04 CAD
CAAA11040-03 CAAA11040-06
Palladium(II) oxide, anhydrous ≥99.9% (metals basis) Pd ≥ 85%
Palladium (II) oxide

Catalytic reduction, in organic synthesis


Purity: 99.9% CAS No.: 1314-08-5 Molecular Formula: PdO Formula Weight: 122.40 Physical Form: -22 Mesh Powder Density: 8.70 Appearance: Brown Odor: Odorless Melting Point: 870° Solubility: Insoluble in water, acids. Slightly soluble in aqua regia Merck Reference: 14,6993 MDL No.: MFCD00011172


Catalytic reduction, in organic synthesis

Formula: OPd
MW: 122.42 g/mol
Melting Pt: 870 °C
Density: 8.70
Storage Temperature: Ambient
MDL Number: MFCD00011172
CAS Number: 1314-08-5
EINECS: 215-218-3
Merck Index: 14,6993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR