Palladium(II) nitrate hydrate (≥39% Pd) ≥99.8% (metals basis) Pd ≥ 39%

Supplier: Thermo Scientific Chemicals
Danger

11035-04 11035-09 11035-18
CAAA11035-04EA 481.9 CAD
CAAA11035-04 CAAA11035-09 CAAA11035-18
Palladium(II) nitrate hydrate (≥39% Pd) ≥99.8% (metals basis) Pd ≥ 39%
Palladium(II) nitrate hydrate

MDL: MFCD00011169 Soluble in water with turbidity, precipitating a brown basic salt. Completely soluble in dilute HNO3


Purity: 99.8% CAS No.: 10102-05-3 Molecular Formula: N2O6Pd.xH2O Formula Weight: 230.43(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Red to brown Melting Point: dec. Solubility: Soluble in water with turbidity, precipitating a brown basic salt. Completely soluble in dilute HNO3 Merck Reference: 14,6992 UN No.: 1477 MDL No.: MFCD00011169

Formula: N₂O₆Pd·H₂O
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD00011169
CAS Number: 313222-87-6
EINECS: 233-265-8
UN: 1477
ADR: 5.1,III
Merck Index: 14,6992

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR