Indium ≥99.99% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
11024-06
11024-14
CAAA11024-06EA
188.02
CAD
CAAA11024-06
CAAA11024-14
Indium ≥99.99% (metals basis), powder -325 mesh
Indium
MDL: MFCD00134048
Purity: 99.99%
CAS No.: 7440-74-6
Molecular Formula: In
Formula Weight: 114.82
Physical Form: Powder
Density: 7.31
Appearance: Silver-grey
Odor: Odorless
Melting Point: 156.6°
Boiling Point: 2072°
UN No.: 3089
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2072 °C Melting Pt: 156.6 °C Density: 7.31 Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...