Calcium fluoride 99%

Supplier: Thermo Scientific Chemicals
Warning

11022-09 11022-18 11022-30
CAAA11022-09EA 91.8 CAD
CAAA11022-09 CAAA11022-18 CAAA11022-30
Calcium fluoride 99%
Calcium fluoride

MDL: MFCD00010907


Purity: 99% CAS No.: 7789-75-5 Molecular Formula: CaF2 Formula Weight: 78.08 Physical Form: Transparent Crystals Density: 3.18 Refractive Index: 1.434 Sensitivity: Hygroscopic Odor: Odorless Melting Point: 1403° Boiling Point: ca 2500° Merck Reference: 14,1667 MDL No.: MFCD00010907

Formula: CaF₂
MW: 78.07 g/mol
Boiling Pt: 2500
Melting Pt: 1418
Density: 3.18
Storage Temperature: Ambient
MDL Number: MFCD00010907
CAS Number: 7789-75-5
EINECS: 232-188-7
Merck Index: 14,1667

Order Now

Specification Test Results

Form Transparent Crystals


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR