Magnesium fluoride ≥99.9% (metals basis), Optical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
11021-09
11021-18
CAAA11021-09EA
72.05
CAD
CAAA11021-09
CAAA11021-18
Magnesium fluoride ≥99.9% (metals basis), Optical Grade
Magnesium fluoride
MDL: MFCD00011108
Purity: 99.9%
CAS No.: 7783-40-6
Molecular Formula: MgF2
Formula Weight: 62.30
Physical Form: Broken Crystals
Density: 3.148
Refractive Index: 1.35-1.38
Appearance: White
Odor: Odorless
Melting Point: 1261°
Merck Reference: 14,5665
MDL No.: MFCD00011108
Formula:
MgF₂ Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1261 °C Density: 3.148 Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 14,5665 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...