Sodium chloride, Optical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Saline, NaCl, Sea salt, Halite

11019-18 11019-30
CAAA11019-18EA 172.22 CAD
CAAA11019-18 CAAA11019-30
Sodium chloride, Optical Grade
Sodium chloride

MDL: MFCD00003477 Fieser: 6,285


CAS No.: 7647-14-5 Molecular Formula: NaCl Formula Weight: 58.44 Physical Form: Broken Crystals Density: 2.165 Refractive Index: 1.5442 Sensitivity: Hygroscopic Appearance: White Odor: Odorless pH: 6-9 Melting Point: 801° Boiling Point: 1413° Merck Reference: 14,8599 MDL No.: MFCD00003477

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
Density: 2.165
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR