Sodium acetate, anhydrous ≥99.997% (metals basis)
Supplier: Thermo Scientific Chemicals
Warning
|
11005-06
11005-14
11005-22
CAAA11005-06EA
70.47
CAD
CAAA11005-06
CAAA11005-14
CAAA11005-22
Sodium acetate, anhydrous ≥99.997% (metals basis)
Sodium acetate
MDL: MFCD00012459
Beilstein Registry No.: 3595639
Fieser: 1,1024 5,59
Purity: 99.997%
CAS No.: 127-09-3
Molecular Formula: NaOOCCH3
Formula Weight: 82.03
Physical Form: Powder
Density: 1.528
pH: 7,5-9
Melting Point: 324°
Beilstein Reference: 3595639
Merck Reference: 14,8571
MDL No.: MFCD00012459
Formula:
H₃CCOONa Boiling Pt: 881.4 °C (1013 hPa) Melting Pt: 324 °C Density: 1.528 Flash Pt: >250 °C (closed cup) Storage Temperature: Ambient |
MDL Number:
MFCD00012459 CAS Number: 127-09-3 EINECS: 204-823-8 REACH: 01-2119485123-42 Merck Index: 14,8571 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...