Sodium fluoride ≥99.99% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
11003-06
11003-14
11003-22
CAAA11003-06EA
58.78
CAD
CAAA11003-06
CAAA11003-14
CAAA11003-22
Sodium fluoride ≥99.99% (metals basis)
Sodium fluoride
MDL: MFCD00003524
Fieser: 1,1073 2,382 6,473
Purity: 99.99%
CAS No.: 7681-49-4
Molecular Formula: NaF
Formula Weight: 41.99
Physical Form: Powder
Density: 2.78
Refractive Index: 1.336
Odor: Odorless
Melting Point: 993°
Boiling Point: 1704°
Merck Reference: 14,8618
UN No.: 1690
MDL No.: MFCD00003524
Formula:
NaF Boiling Pt: 1704 °C Melting Pt: 993 °C Density: 2.78 Storage Temperature: Ambient |
MDL Number:
MFCD00003524 CAS Number: 7681-49-4 EINECS: 231-667-8 UN: 1690 ADR: 6.1,III Merck Index: 14,8618 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...