Zinc selenide ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Zinc (II) selenide

10999-09 10999-18 10999-30
CAAA10999-09EA 134.14 CAD
CAAA10999-09 CAAA10999-18 CAAA10999-30
Zinc selenide ≥99.999% (metals basis)
Zinc selenide

MDL: MFCD00011296


Purity: 99.999% CAS No.: 1315-09-9 Molecular Formula: ZnSe Formula Weight: 144.33 Physical Form: Powder Density: 5.42 Melting Point: 1100° Merck Reference: 14,10156 UN No.: 3283 MDL No.: MFCD00011296

Formula: SeZn
MW: 144.35 g/mol
Melting Pt: >1100 °C
Density: 5.42
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 14,10156

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR