Calcium fluoride ≥99.95% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
10998-14
10998-22
CAAA10998-14EA
130.82
CAD
CAAA10998-14
CAAA10998-22
Calcium fluoride ≥99.95% (metals basis)
Calcium fluoride
MDL: MFCD00010907
Purity: 99.95%
CAS No.: 7789-75-5
Molecular Formula: CaF2
Formula Weight: 78.08
Physical Form: -325 Mesh Powder
Density: 3.18
Refractive Index: 1.434
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 1403°
Boiling Point: ca 2500°
Merck Reference: 14,1667
MDL No.: MFCD00010907
Formula:
CaF₂ Boiling Pt: ∼2500 °C Melting Pt: 1403 °C Density: 3.18 Storage Temperature: Ambient |
MDL Number:
MFCD00010907 CAS Number: 7789-75-5 EINECS: 232-188-7 Merck Index: 14,1667 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...