Calcium carbonate ≥99.95% (metals basis)

Supplier: Thermo Scientific Chemicals

10996-14 10996-22
CAAA10996-14EA 153.1 CAD
CAAA10996-14 CAAA10996-22
Calcium carbonate ≥99.95% (metals basis)
Calcium carbonate

MDL: MFCD00010906


Purity: 99.95% CAS No.: 471-34-1 Molecular Formula: CaCO3 Formula Weight: 100.09 Physical Form: Powder Density: 2.930 Refractive Index: 1.6583 Odor: Odorless Melting Point: 800° dec. Merck Reference: 14,1657 MDL No.: MFCD00010906

Formula: CaCO₃
Melting Pt: 800 °C (dec.)
Density: 2.930
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 14,1657

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR