Barium chloride, anhydrous ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10995-02
10995-06
10995-14
10995-22
CAAA10995-02EA
49.77
CAD
CAAA10995-02
CAAA10995-06
CAAA10995-14
CAAA10995-22
Barium chloride, anhydrous ≥99.998% (metals basis)
Barium chloride
MDL: MFCD00003445
Purity: 99.998%
CAS No.: 10361-37-2
Molecular Formula: BaCl2
Formula Weight: 208.25
Physical Form: Powder, may contain small lumps
Density: 3.9
Appearance: White
Odor: Odorless
Melting Point: 963°
Boiling Point: 1560°
Merck Reference: 14,971
UN No.: 1564
MDL No.: MFCD00003445
Formula:
BaCl₂ Boiling Pt: 1560 °C Melting Pt: 963 °C Density: 3.9 Storage Temperature: Ambient |
MDL Number:
MFCD00003445 CAS Number: 10361-37-2 EINECS: 233-788-1 UN: 1564 ADR: 6.1,III Merck Index: 14,971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...