Acetic acid glacial ≥99.9985% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10994-AC
10994-AE
10994-AP
CAAA10994-ACEA
66.52
CAD
CAAA10994-AC
CAAA10994-AE
CAAA10994-AP
Acetic acid glacial ≥99.9985% (metals basis)
Acetic acid
MDL: MFCD00036152
Fieser: 2,5 5,3 7,1 8,1 19,1 21,1
Purity: 99.9985%
CAS No.: 64-19-7
Molecular Formula: CH3CO2H
Formula Weight: 60.05
Flash Point: 40°(104°F)
Physical Form: Liquid
Density: 1.049
Refractive Index: 1.3721
Odor: Pungent
pH: 2,5
Melting Point: 16.6°
Boiling Point: 118.1°
Merck Reference: 14,55
UN No.: 2789
MDL No.: MFCD00036152
Formula:
H₃CCOOH Boiling Pt: 118.1 °C Melting Pt: 16.6 °C Density: 1.049 Flash Pt: 40 °C (104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036152 CAS Number: 64-19-7 EINECS: 200-580-7 UN: 2789 ADR: 8,II REACH: 01-2119475328-30 Merck Index: 14,55 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...