Hydrobromic acid 48% (w/w) in aqueous solution , 99,9999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

10991-AA 10991-AE 10991-AP
CAAAA10991-AAEA 77.89 CAD
CAAAA10991-AA CAAA10991-AE CAAA10991-AP
Hydrobromic acid 48% (w/w) in aqueous solution , 99,9999% (metals basis)
Hydrogen bromide

MDL: MFCD00011323 Beilstein Registry No.: 3587158 Fieser: 1,450 2,214 3,154 18,180


Purity: 99.9999% CAS No.: 10035-10-6 Molecular Formula: HBr Formula Weight: 80.92 Physical Form: Liquid Density: 1.490 Appearance: Colorless to faint yellow Beilstein Reference: 3587158 Merck Reference: 14,4778 UN No.: 1788 MDL No.: MFCD00011323

Formula: HBr
Density: 1.490
Storage Temperature: Ambient
MDL Number: MFCD00011323
CAS Number: 10035-10-6
EINECS: 233-113-0
UN: 1788
ADR: 8,II
Merck Index: 14,4778

Order Now

Specification Test Results

Form Liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR