Nitric acid 65 - 70% ≥99.9999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nitric acid solution
10984-AC
10984-AK
10984-AP
CAAA10984-ACEA
124.5
CAD
CAAA10984-AC
CAAA10984-AK
CAAA10984-AP
Nitric acid 65 - 70% ≥99.9999% (metals basis)
Nitric acid
MDL: MFCD00011349
Purity: 99.9999%
CAS No.: 7697-37-2
Molecular Formula: HNO3
Formula Weight: 63.01
Physical Form: Liquid
Density: 1.41
Appearance: Colorless to pale yellow
Odor: Pungent
Boiling Point: 121°
Merck Reference: 14,6577
UN No.: 2031
MDL No.: MFCD00011349
Formula:
HNO₃ MW: 63.01 Boiling Pt: 121 °C Melting Pt: –42 °C Density: 1.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8 II Merck Index: 14,6577 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...