Perchloric acid ≥70% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
10983-AA
10983-AD
10983-AK
CAAAA10983-AAEA
83.11
CAD
CAAAA10983-AA
CAAA10983-AD
CAAA10983-AK
Perchloric acid ≥70% (metals basis)
Perchloric acid
MDL: MFCD00011325
Fieser: 1,796 2,309 3,220 5,506 6,453 7,279 8,386 11,402 12,402
Purity: 99.9985%
CAS No.: 7601-90-3
Molecular Formula: HClO4
Formula Weight: 100.46
Flash Point: 113°(235°F)
Physical Form: Liquid
Density: 1.670
Appearance: Colorless
Odor: Odorless
Melting Point: -18°
Boiling Point: 203°
Merck Reference: 14,7153
UN No.: 1873
MDL No.: MFCD00011325
Formula:
HClO₄ Boiling Pt: 203 °C Melting Pt: –18 °C Density: 1.670 Flash Pt: 113 °C (235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1,I Merck Index: 14,7153 |
|
Specification Test Results
Form | Liquid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...