Potassium bromide ≥99.92% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

10982-14 10982-22
CAAA10982-14EA 158 CAD
CAAA10982-14 CAAA10982-22
Potassium bromide ≥99.92% (metals basis)
Potassium bromide

MDL: MFCD00011358 Fieser: 21,358


Purity: 99.92% CAS No.: 7758-02-3 Molecular Formula: KBr Formula Weight: 119.01 Physical Form: Crystalline Density: 2.75 Refractive Index: 1.559 Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 730° Boiling Point: 1435° Merck Reference: 14,7618 MDL No.: MFCD00011358

Formula: KBr
Boiling Pt: 1435 °C
Melting Pt: 730 °C
Density: 2.75
Storage Temperature: Ambient
MDL Number: MFCD00011358
CAS Number: 7758-02-3
EINECS: 231-830-3
Merck Index: 14,7618

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR