Potassium carbonate sesquihydrate 98.5-101.0% ACS

Supplier: Thermo Scientific Chemicals
Warning

10981-22 10981-36
CAAA10981-22EA 53.72 CAD
CAAA10981-22 CAAA10981-36
Potassium carbonate sesquihydrate 98.5-101.0% ACS
Potassium carbonate sesquihydrate

Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent


Purity: 98.5-101.0% CAS No.: 6381-79-9 Molecular Formula: K2CO3.1.5H2O Formula Weight: 165.23 (138.21anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Solubility: Soluble in water. Practically insoluble in alcohol Merck Reference: 14,7670 MDL No.: MFCD00149925


Manufacturing of soaps, pigments, printing ink, glass, pottery, other potassium salts, as an analytical lab reagent

Formula: CK₂O₃·1,5H₂O
MW: 165.23 g/mol
Melting Pt: 891
Density: 2.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3
Merck Index: 14,7670

Order Now

Specification Test Results

Insoluble matter 0.01%
Cl 0.3%
PO7 0.001%
SiO 0.005%
Sulfur compounds (as SO3) 0.004%
Ca 0.005%
MG 0.002%
Heavy metals (as Pb) 5 ppm
Fe 5 ppm
NA 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR