Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

10975-BW 10975-BZ 10975-CC
CAAA10975-BWEA 218.04 CAD
CAAA10975-BW CAAA10975-BZ CAAA10975-CC
Copper ≥99.999% (metals basis), wire, Puratronic®, Ø 2.0 mm (0.08 in)
Copper

MDL: MFCD00010965


Purity: 99.999% CAS No.: 7440-50-8 Physical Form: ≈28.1g/m Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR