Copper ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

10972-G6 10972-H2
CAAA10972-G6EA 172.22 CAD
CAAA10972-G6 CAAA10972-H2
Copper ≥99.9985% (metals basis), wire, Puratronic®, Ø 0.25 mm (0.01 in)
Copper

MDL: MFCD00010965


Purity: 99.9985% CAS No.: 7440-50-8 Physical Form: ≈0.44g/m Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
Boiling Pt: 2595 °C (1013 hPa)
Melting Pt: 1083 °C
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR